Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-N'-oleoylethylenediamine 97.0+%, TCI America™
SDP

Catalog No. D051725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D051725G 25 g
D05175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D051725G Supplier TCI America Supplier No. D051725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13282-67-2
Molecular Formula C24H48N2O
Molecular Weight (g/mol) 380.66
MDL Number MFCD00027344
InChI Key WCSBDRIEINNXNW-BUHFOSPRSA-N
PubChem CID 6146519
IUPAC Name (9E)-N-[2-(diethylamino)ethyl]octadec-9-enamide
SMILES CCCCCCCC\C=C\CCCCCCCC(=O)NCCN(CC)CC

Specifications

Color Brown-Yellow
Quantity 25 g
Synonym N′-Oleoyl-N,N-diethylethylenediamine
Formula Weight 380.66
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-N′-oleoylethylenediamine
EINECSNumber (9)-0276
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.