Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylacetamide, 99%

Catalog No. AAB2076222
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2076222 100 g
AAB2076214 25 g
AAB2076236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2076222 Supplier Thermo Scientific Chemicals Supplier No. B2076222
Only null left
Add to Cart

CAS: 685-91-6 | C6H13NO | 115.176 g/mol

N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685-91-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00009047
InChI Key AJFDBNQQDYLMJN-UHFFFAOYSA-N
Synonym diethylacetamide, acetamide, n,n-diethyl, n-acetyldiethylamine, unii-6yo81d7st6, n,n-diaethylacetamid german, n,n-diethyl-acetamide, acetic acid, amide, n,n-diethyl, acetamide,n,n-diethyl, n,n-diaethylacetamid, n,n-diethylacetoamide
PubChem CID 12703
IUPAC Name N,N-diethylacetamide
SMILES CCN(CC)C(=O)C

Specifications

Density 0.92
Boiling Point 182°C to 186°C
Flash Point 76°C (169°F)
Refractive Index 1.44
Quantity 100 g
Beilstein 1209428
Solubility Information Soluble in water.
Formula Weight 115.18
Percent Purity 99%
Chemical Name or Material N,N-Diethylacetamide
Hazard Category H227-H373
Hazard Statement GHS H Statement
H302-H315-H319-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P260-P280-P314-P370+P378q-P501c
EINECSNumber 211-685-2
RTECSNumber AB7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.