Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylacrylamide (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. D373725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D373725G 25 g
D3737500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D373725G Supplier TCI America Supplier No. D373725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2675-94-7
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00078408
InChI Key OVHHHVAVHBHXAK-UHFFFAOYSA-N
PubChem CID 17583
IUPAC Name N,N-diethylprop-2-enamide
SMILES CCN(CC)C(=O)C=C

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 g
Formula Weight 127.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylacrylamide (stabilized with MEHQ)
EINECSNumber (2)-1017
RTECSNumber AS3471000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.