Learn More
N,N-Diethylaniline, 99%
Description
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.
Solubility
Miscible with water and acetone. Slightly miscible with chloroform, alcohol and ether.
Notes
Incompatible with strong oxidizing agents and strong acids. .
Specifications
Specifications
| Melting Point | -38°C |
| Density | 0.936 |
| Boiling Point | 215°C to 216°C |
| Flash Point | 85°C (185°F) |
| Refractive Index | 1.542 |
| Quantity | 2500 mL |
| UN Number | UN2432 |
| Beilstein | 742483 |
| Merck Index | 14,3114 |
| Solubility Information | Miscible with water and acetone. Slightly miscible with chloroform,alcohol and ether. |
| Show More |
Safety and Handling
| Hazard Category | H227-H301+H311+H331-H373 |
| Hazard Statement | GHS H Statement H301-H311-H331-H373-H227 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Combustible liquid. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N,N DIETHYLANILINE |
| EINECSNumber | 202-088-8 |
| RTECSNumber | BX3400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.