Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diethylethylenediamine 95.0+%, TCI America™
SDP

Catalog No. D028725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D028725ML 25 mL
D02875ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D028725ML Supplier TCI America Supplier No. D028725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111-74-0
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00009033
InChI Key CJKRXEBLWJVYJD-UHFFFAOYSA-N
Synonym n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide
PubChem CID 67105
ChEBI CHEBI:182290
IUPAC Name ethyl[2-(ethylamino)ethyl]amine
SMILES CCNCCNCC

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 mL
UN Number 2685
Formula Weight 116.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N′-Diethylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.