Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylformamide, 99%

Catalog No. p-109182
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181370250 25 mL Glass bottle
AC181371000 100 mL Glass bottle
AC181375000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC181370250 Supplier Thermo Scientific Chemicals Supplier No. 181370250
Only null left
Add to Cart

CAS: 617-84-5 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-84-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003287
InChI Key SUAKHGWARZSWIH-UHFFFAOYSA-N
Synonym diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
PubChem CID 12051
IUPAC Name N,N-diethylformamide
SMILES CCN(CC)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 176.0°C to 177.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Linear Formula HCON(C2H5)2
Quantity 25 mL
Beilstein 04, 109
Specific Gravity 0.9
Formula Weight 101.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Diethylformamide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get med
EINECSNumber 210-533-2
RTECSNumber LQ1925000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.