Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylglycine Sodium Salt 97.0+%, TCI America™
SDP

Catalog No. D0509100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0509100G 100 g
D050925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0509100G Supplier TCI America Supplier No. D0509100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5426-55-1
Molecular Formula C6H13NNaO2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00053497
InChI Key XZQDOIKSUNQDGV-UHFFFAOYSA-N
PubChem CID 23670834
IUPAC Name 2-(diethylamino)acetic acid sodium
SMILES [Na].CCN(CC)CC(O)=O

Specifications

Color White-Yellow
Quantity 100 g
Formula Weight 153.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diethylglycine Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.