Learn More
Cayman Chemical N N-DIferuloylputrescIn 1mg
SDP
Supplier:  Cayman Chemical 414361 MG
Encompass_Preferred
A SERT-nNOS protein-protein interaction inhibitor increases in vivo serotonergic neuron firing diminished by decreased SERT levels in SERT-Cre transgenic mice which have reduced SERT levels at 10 mg/kg reduces immobility time in the tail suspension test and forced swim test in mice at 20 and 40 mg/kg
Catalog No. 50-294-4259
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.