Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropylethylamine, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611851 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
200 L
Packaging:
Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-851 100 mL Drum
18-611-852 500 mL Drum
18-611-850 4 L Drum
18-611-849 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-851 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D2163100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7087-68-5

Specifications

CAS Max % 1
Boiling Point 127°C / 260.6°F
Packaging Drum
Quantity 100 mL
Molecular Weight (g/mol) 129.25
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material N,N-Diisopropylethylamine

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.