Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropylethylamine trihydrofluoride, 90%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Plastic bottle
131600-43-6
C8H193HF
189.26
MFCD00144880
AJRRXKJZYYBJPY-UHFFFAOYSA-N
diisopropylethylamine trihydrofluoride,n-ethyl-n-isopropylpropan-2-amine trihydrofluoride,n,n-diisopropylethylamine trihydrofluoride,acmc-1bx1e,ksc490i4d,n,n,-diisopropylethylamine trihydrofluoride,n-ethyl-n-isopropylpropan-2-aminetrihydrofluoride,n-ethyl-n-isopropyl-propan-2-amine trihydrofluoride,n-ethyl-n-propan-2-yl-2-propanamine trihydrofluoride,n-ethyl-n-propan-2-yl-propan-2-amine trihydrofluoride
45108321
N-ethyl-N-propan-2-ylpropan-2-amine;trihydrofluoride
CCN(C(C)C)C(C)C.F.F.F
131600-43-6
C8H193HF
189.26
MFCD00144880
AJRRXKJZYYBJPY-UHFFFAOYSA-N
diisopropylethylamine trihydrofluoride,n-ethyl-n-isopropylpropan-2-amine trihydrofluoride,n,n-diisopropylethylamine trihydrofluoride,acmc-1bx1e,ksc490i4d,n,n,-diisopropylethylamine trihydrofluoride,n-ethyl-n-isopropylpropan-2-aminetrihydrofluoride,n-ethyl-n-isopropyl-propan-2-amine trihydrofluoride,n-ethyl-n-propan-2-yl-2-propanamine trihydrofluoride,n-ethyl-n-propan-2-yl-propan-2-amine trihydrofluoride
45108321
N-ethyl-N-propan-2-ylpropan-2-amine;trihydrofluoride
CCN(C(C)C)C(C)C.F.F.F
Boiling Point 93°C to 95°C (4.0mmHg)
Flash Point 40.5°C
Specific Gravity 0.965
Density 0.9650g/mL
Infrared Spectrum Authentic
Color Undesignated
Packaging Plastic bottle
Quantity 1g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 90%
Solubility Information Solubility in water: soluble
Formula Weight 189.26
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material N, N-Diisopropylethylamine trihydrofluoride
Show More Show Less

GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
Fatal if swallowed.
Flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Rinse mouth.
Immediately call a POISON CENT

GHS Signal Word: Danger