Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diisopropylethylenediamine 97.0+%, TCI America™
SDP

Catalog No. D392325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D392325G 25 g
D39235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D392325G Supplier TCI America Supplier No. D392325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4013-94-9
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00008864
InChI Key MFIGJRRHGZYPDD-UHFFFAOYSA-N
Synonym n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine
PubChem CID 77628
IUPAC Name (propan-2-yl)({2-[(propan-2-yl)amino]ethyl})amine
SMILES CC(C)NCCNC(C)C

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 g
UN Number 2733
Formula Weight 144.26
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Diisopropylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.