Learn More
N,N-Dimethyl-n-octylamine, 95%
Description
N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide
Solubility
Slightly miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and copper.
Specifications
Specifications
| Melting Point | -57°C |
| Density | 0.765 |
| Boiling Point | 195°C |
| Flash Point | 65°C (149°F) |
| Refractive Index | 1.4243 |
| Quantity | 50 g |
| UN Number | UN2922 |
| Beilstein | 1698081 |
| Sensitivity | Air Sensitive |
| Solubility Information | Slightly miscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H227-H301-H314 |
| Hazard Statement | GHS H Statement H301-H314-H318-H227 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 230-939-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.