Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-p-phenylenediamine, 96%

Catalog No. AAA159620B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA159620B 1000 g
AAA1596218 50 g
AAA1596230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA159620B Supplier Thermo Scientific Chemicals Supplier No. A159620B
Add to Cart

CAS: 99-98-9 | C8H12N2 | 136.20 g/mol

N,N-Dimethyl-p-phenylenediamine is a redox indicator. Readily forms stable, red radical cation, involved in a variety of redox reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-phenylenediamine is a redox indicator. Readily forms stable, red radical cation, involved in a variety of redox reactions.

Solubility
Soluble in Water (11 g/L at20°C), diethyl ether, petroleum ether, alcohol and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light and air sensitive. Store under inert gas. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-98-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00007860
InChI Key BZORFPDSXLZWJF-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine, n,n-dimethyl-1,4-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-aminodimethylaniline, dmpd, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, p-amino-n,n-dimethylaniline, 4-amino-n,n-dimethylaniline, p-dimethylamino aniline
PubChem CID 7472
ChEBI CHEBI:15783
IUPAC Name 4-N,4-N-dimethylbenzene-1,4-diamine
SMILES CN(C)C1=CC=C(N)C=C1

Specifications

Melting Point 34°C to 38°C
Density 1.09
Boiling Point 262°C
Flash Point 130°C (266°F)
Quantity 1000 g
UN Number UN2811
Beilstein 508105
Merck Index 14,3254
Solubility Information Soluble in Water (11g/L at20°C),diethyl ether,petroleum ether,alcohol and chloroform.
Formula Weight 136.2
Percent Purity 96%
Chemical Name or Material N,N-Dimethyl-p-phenylenediamine
Show More Show Less
Hazard Category H300-H311+H331-H335-H373
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-807-5
RTECSNumber ST0874000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.