Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-p-Toluidine, Spectrum™ Chemical
SDP

Catalog No. 18620634 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
17.2 kg
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-634 200 L
18-620-631 25 g
18-620-632 100 g
18-620-633 17.2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-634 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D2335200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Synonym: N,N,4-Trimethylaniline, Dimethyl-p-Toluidine, N,N,4-Trimethyl-Benzenamine

N,N-Dimethyl-p-toluidine is used to make acrylic resins and denture materials. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 99-97-8

Specifications

Boiling Point 211°C
Packaging Drum
Quantity 200 L
Percent Purity 98.5%
Physical Form Liquid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.