accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3

CAS RN 99-97-8

IUPAC Name: N,N,4-trimethylaniline
Synonyms: dimethyltolylamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: GYVGXEWAOAAJEU-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C)C=C1
Boiling Point 
  • (3)
  • (6)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (1)
  • (4)
  • (2)
  • (5)
  • (3)
  • (6)
  • (1)
  • (2)
  • (2)
  • (4)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (8)