Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018782 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL

Catalog No. 11-101-8782

Supplier: MilliporeSigma Supelco 723365MLF

Only null left
Add to Cart
Add to Cart

N,N-Dimethylacetamide is a highly polar solvent, which can find applications in a variety of chemical reactions. A mixture of lithium chloride in N,N-dimethylacetamide may be used for the dissolution of cellulose to form a homogeneous solution.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 914°F
CAS 127-19-5
Vapor Pressure 2 mmHg (25°C)
Vapor Density 3 (vs. air)
Density 0.937 g/mL (at 25°C)
Flash Point 64°C - Closed Cup
Physical Form Neat
Linear Formula CH3 CON(CH3)2
MDL Number MFCD00008686
Melting Point -20°C
Boiling Point 164.5°C to 166°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 87.12
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.