Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylethanolamine, 99%

Catalog No. AC116180010
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
25 L
Packaging:
Glass bottle
Metal Drum
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116180010 1 L Glass bottle
AC116182500 250 mL Glass bottle
AC116180051 5 L Plastic drum
AC116180250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC116180010 Supplier Thermo Scientific Chemicals Supplier No. 116180010
Only null left
Add to Cart

CAS: 108-01-0 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-01-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00002846
InChI Key UEEJHVSXFDXPFK-UHFFFAOYSA-N
Synonym 2-dimethylamino ethanol, n,n-dimethylethanolamine, deanol, dimethylaminoethanol, dimethylethanolamine, norcholine, dmae, dmea, bimanol, liparon
PubChem CID 7902
ChEBI CHEBI:271436
IUPAC Name 2-(dimethylamino)ethanol
SMILES CN(C)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.8880g/mL
Boiling Point 139.0°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.4300
Linear Formula HOCH2CH2N(CH3)2
Quantity 1 L
Beilstein 04, 276
Specific Gravity 0.888
Merck Index 15, 2845
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol and ether
Viscosity 3.8 mPa.s (20°C)
Formula Weight 89.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylethanolamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
EINECSNumber 203-542-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.