Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, 99.8%, for spectroscopy

Catalog No. AC167790010
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167790010 1 L Glass bottle
AC167795000 500 mL Glass bottle
AC167790025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC167790010 Supplier Thermo Scientific Chemicals Supplier No. 167790010
Add to Cart
This item is not returnable. View return policy

CAS: 68-12-2 | C3H7NO | 73.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Alkalinity 0.001 meq/g max.
Acidity 0.001 meq/g max.
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 340nm, 0.05 max. at 310nm, 0.1 max. at 295nm, 0.3 max. at 275nm, 1 max. at 270nm
CAS Min % 99.75
CAS Max % 100.0
Melting Point -61.0°C
Density 0.9450g/mL
Boiling Point 153.0°C
Flash Point 58°C
Assay Percent Range 99.75% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.4320
Linear Formula HCON(CH3)2
Quantity 1 L
Residue after Evaporation 0.0015% max.
Beilstein 04, 58
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Specific Gravity 0.945
Merck Index 15, 3267
Specific Rotation 0.94
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Viscosity 0.8 mPa.s (20°C)
Formula Weight 73.09
Percent Purity 99.8%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wea
EINECSNumber 200-679-5
RTECSNumber LQ2100000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.