Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, ACS Reagent (DMF), Reagents

Catalog No. 01338007
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Glass Bottle
Poly Bottle
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-338-007 4 L Glass Bottle
01-338-009 20 L Steel Pail
01-338-008 4 L Poly Bottle
01-338-006 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-338-007 Supplier Reagents Holdings Llc Supplier No. C11962004C
Add to Cart
Add to Cart

N,N-Dimethylformamide, ACS Reagent (DMF), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym Dimethylformamide, DMF, N,N-Dimethylmethanamide
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -60.5°C
Color Colorless to Slightly Yellow
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.944
Boiling Point 152°C
Grade ACS
Chemical Name or Material N,N-Dimethylformamide, ACS Reagent (DMF)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.