Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, Anhydrous, septum-sealed, >99.9%, Solstice

Catalog No. 6000994 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 100 mL
6 x 1 L
Packaging:
Septum-sealed Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-009-94 6 x 100 mL Septum-sealed Bottle
60-009-95 6 x 1 L Septum-sealed Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-009-94 Supplier Solstice Supplier No. AS0760100

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

CAS Max % 100.0000%
Density 0.949 g/cm3
Quantity 6 x 100 mL
Flash Point 58°C
Packaging Septum-sealed Bottle
Physical Form anhydrous
UN Number UN2265
Water 50 ppm max.
Beilstein 605365
Boiling Point 153°C
Solubility Information Miscible in Water at 20°C
Formula Weight 73.10
Percent Purity >99.9%
Chemical Name or Material N,N-Dimethylformamide, Anhydrous, septum-sealed, for applications requiring low water content, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: N,N-Dimethylformamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.