Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, Biotech Grade, ≥99.9%,Solstice

Catalog No. 60046986 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2 L
6 x 1 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-986 1 L Glass Bottle
60-046-987 2 L Glass Bottle
60-046-988 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-986 Supplier Solstice Supplier No. 4944881L
Add to Cart
Add to Cart

Biotech Grade,≥99.9%

  • Solstice developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Solstice continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 7 (at 20°C)
Melting Point -61°C
Color Colorless
Density 0.944g/mL
Quantity 1 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Odor Weak
Packaging Glass Bottle
Physical Form Liquid
Linear Formula HCON(CH3)2
Vapor Pressure 2.7mmHg (20°C)
UN Number UN2265
Beilstein 605365
Boiling Point 152°C to 153°C
Formula Weight 73.09g/mol
Percent Purity ≥99.9%
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Statement P201-P280-P305 + P351 + P338-P308 + P313
DOTInformation 3/PGIII
RTECSNumber 200-679-5
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.