Learn More
N,N-Dimethylformamide dimethyl acetal, >95%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Density | 0.897 g/mL (at 25°C) |
| CAS | 4637-24-5 |
| Boiling Point | 102°C to 103°C (720 mmHg) |
| Flash Point | 5°C - Closed Cup |
| Packaging | Ampule |
| Linear Formula | (CH3)2 NCH(OCH3)2 |
| Quantity | 25 mL |
| Formula Weight | 119.16 |
| Percent Purity | >95% |
| Grade | For GC derivatization |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.