Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide Dineopentyl Acetal 96.0+%, TCI America™
SDP

Catalog No. D159525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D159525ML 25 mL
D15955ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D159525ML Supplier TCI America Supplier No. D159525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4909-78-8
Molecular Formula C13H30NO2
Molecular Weight (g/mol) 232.39
MDL Number MFCD00008851
InChI Key KEXFRBIOHPDZQM-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dineopentyl acetal, methanamine, 1,1-bis 2,2-dimethylpropoxy-n,n-dimethyl, n,n-dimethyl-1,1-bis neopentyloxy methanamine, dimethylformamide dineopentyl acetal, 1,1-bis 2,2-dimethylpropoxy-n,n-dimethylmethanamine, bis 2,2-dimethylpropoxy methyl dimethylamine, acmc-1al8s, 1,1-bis 2,2-dimethylpropoxy-n,n,n-trimethylamine, 1,1-dineopentyloxytrimethylamine, dimethylformamide dineopentylacetal
PubChem CID 78623
IUPAC Name [bis(2,2-dimethylpropoxy)methyl]dimethylazanium
SMILES C[NH+](C)C(OCC(C)(C)C)OCC(C)(C)C

Specifications

Color Colorless
Boiling Point 87°C
Quantity 25 mL
Formula Weight 231.38
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Dineopentyl Acetal [for Esterification]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.