Learn More
N,N-Dimethylformamide, HPLC Grade, 99.7+%
Description
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is used as a mobile phase in High Performance Liquid Chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 61°C |
| Color | Colorless |
| Appearance | Colorless liquid |
| Density | 0.944 |
| Quantity | 4 x 4 L |
| Flash Point | 57°C (135°F) |
| Odor | Amine-like |
| Physical Form | Liquid |
| Refractive Index | 1.431 |
| Assay Percent Range | >99.7% |
| Show More |
Safety and Handling
| Hazard Category | H226-H312+H332-H319-H335-H336-H350-H360D |
| Hazard Statement | Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 3; Packing Group: III |
| EINECSNumber | 200-679-5 |
| RTECSNumber | LQ2100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.