Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, Purified (DMF), Reagents

Catalog No. 01337281
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-281 20 L
01-337-283 1 L
01-337-280 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-281 Supplier Reagents Holdings Llc Supplier No. C219620020E6
Add to Cart
Add to Cart

N,N-Dimethylformamide, Purified (DMF), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym Dimethylformamide, DMF, N,N-Dimethylmethanamide
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -60.5°C
Color Colorless to Slight Yellow
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.944
Boiling Point 152°C to 153°C
Grade Pure
Chemical Name or Material N,N-Dimethylformamide, Purified (DMF)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.