Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, Reagent Grade, ≥99%, Solstice

Catalog No. 60047936 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 L
4 x 2.5 L
6 x 1 L
Packaging:
Poly Bottle
Returnable Stainless Steel
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-936 4 x 2.5 L Poly Bottle
60-047-934 1 L Poly Bottle
60-047-935 2.5 L Poly Bottle
60-047-939 6 x 1 L Poly Bottle
60-048-509 20 L Returnable Stainless Steel
60-047-938 5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-047-936 Supplier Solstice Supplier No. D1585504X2.5L
Add to Cart
Add to Cart

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.944 g/mL(lit.)
Quantity 4 x 2.5 L
Explosion Limit 0.152
Flash Point 57.5°C
Ignition Point 833°F
Packaging Poly Bottle
Linear Formula HCON(CH3)2
Vapor Pressure 2.7 mmHg (20°C)
Water max. 0.1%
UN Number UN2265
Beilstein 605365
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
Boiling Point 152°C to 153°C
Formula Weight 73.10
Percent Purity >99%
Grade Reagent
Chemical Name or Material N,N-Dimethylformamide, Reagent Grade, ≥99%
Show More Show Less
Hazard Statement P201-P210-P261-P280-P308 + P313-P370 + P378
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: N,N-Dimethylformamide
EINECSNumber 200-679-5
RTECSNumber LQ2100000
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.