Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylpropargylamine 98.0+%, TCI America™
SDP

Catalog No. D279425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D279425ML 25 mL
D27945ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D279425ML Supplier TCI America Supplier No. D279425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7223-38-3
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00008575
InChI Key ILBIXZPOMJFOJP-UHFFFAOYSA-N
Synonym 3-dimethylamino-1-propyne, 1-dimethylamino-2-propyne, 2-propyn-1-amine, n,n-dimethyl, n,n-dimethylpropargylamine, dimethyl prop-2-yn-1-yl amine, n,n-dimethyl-2-propyn-1-amine, dimethyl prop-2-ynyl amine, n,n-dimethyl propargylamine, n,n-dimethyl-2-propynylamine, dimethylpropargylamine
PubChem CID 81643
IUPAC Name N,N-dimethylprop-2-yn-1-amine
SMILES CN(C)CC#C

Specifications

Color Yellow
Quantity 25 mL
UN Number 2733
Formula Weight 83.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylpropargylamine
EINECSNumber (2)-0144
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.