Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dimethylquinacridone 96.0+%, TCI America™
SDP

Catalog No. D26871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26871G 1 g
D26875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26871G Supplier TCI America Supplier No. D26871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19205-19-7
Molecular Formula C22H16N2O2
Molecular Weight (g/mol) 340.382
MDL Number MFCD00683078
InChI Key SCZWJXTUYYSKGF-UHFFFAOYSA-N
Synonym 5,12-Dihydro-5,12-dimethylquino[2,3-b]acridine-7,14-dione, DMQA
PubChem CID 2844779
IUPAC Name 5,12-dimethylquinolino[2,3-b]acridine-7,14-dione
SMILES CN1C2=CC=CC=C2C(=O)C3=CC4=C(C=C31)C(=O)C5=CC=CC=C5N4C

Specifications

Quantity 1 g
Formula Weight 340.38
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dimethylquinacridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.