Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dioctadecylmethylamine 97.0+%, TCI America™
SDP

Catalog No. D22295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22295G 5 g
D222925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22295G Supplier TCI America Supplier No. D22295G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4088-22-6 | Formula: C37H77N | Weight: 536.03 g/mol

Chemical Identifiers

CAS 4088-22-6
Molecular Formula C37H77N
Molecular Weight (g/mol) 536.03
MDL Number MFCD00075495
InChI Key VFLWKHBYVIUAMP-UHFFFAOYSA-N
Synonym N-Methyldioctadecylamine
PubChem CID 77709
IUPAC Name methyldioctadecylamine
SMILES CCCCCCCCCCCCCCCCCCN(C)CCCCCCCCCCCCCCCCCC

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 5 g
Formula Weight 536.03
Percent Purity ≥97.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dioctadecylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.