Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diphenyl-N,N'-bis(p-tolyl)-1,4-phenylenediamine 97.0+%, TCI America™
SDP

Catalog No. D49051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49051G 1 g
D49055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49051G Supplier TCI America Supplier No. D49051G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138171-14-9
Molecular Formula C32H28N2
Molecular Weight (g/mol) 440.59
MDL Number MFCD01310665
InChI Key FQNVFRPAQRVHKO-UHFFFAOYSA-N
PubChem CID 16218148
IUPAC Name 1-N,4-N-bis(4-methylphenyl)-1-N,4-N-diphenylbenzene-1,4-diamine
SMILES CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=C(C=C5)C

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 1 g
Synonym 4′C,4′C′C′C-Dimethyl-N,N,N′C,N′C-tetraphenyl-1,4-phenylenediamine
Formula Weight 440.59
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenyl-N,N′-bis(p-tolyl)-1,4-phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.