Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diphenyl-N,N'-di(p-tolyl)benzidine 98.0+%, TCI America™
SDP

Catalog No. D48341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48341G 1 g
D4834200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48341G Supplier TCI America Supplier No. D48341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20441-06-9
Molecular Formula C38H32N2
Molecular Weight (g/mol) 516.688
MDL Number MFCD00799301
InChI Key UNZWWPCQEYRCMU-UHFFFAOYSA-N
Synonym N,N′C-Bis(4-methylphenyl)-N,N′C-diphenylbenzidine, 4,4′C-Bis[N-phenyl-N-(p-tolyl)amino]biphenyl
PubChem CID 640095
IUPAC Name 4-methyl-N-[4-[4-(N-(4-methylphenyl)anilino)phenyl]phenyl]-N-phenylaniline
SMILES CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=CC=C(C=C6)C

Specifications

Melting Point 167°C
Color White-Yellow
Quantity 1 g
Formula Weight 516.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenyl-N,N′-di(p-tolyl)benzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.