Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diphenylformamide 98.0+%, TCI America™
SDP

Catalog No. D089025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D089025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D089025G Supplier TCI America Supplier No. D089025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 607-00-1
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.237
MDL Number MFCD00003282
InChI Key DCNUQRBLZWSGAV-UHFFFAOYSA-N
Synonym Formyldiphenylamine
PubChem CID 69081
IUPAC Name N,N-diphenylformamide
SMILES C1=CC=C(C=C1)N(C=O)C2=CC=CC=C2

Specifications

Melting Point 72°C
Boiling Point 337°C
Quantity 25 g
Formula Weight 197.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diphenylformamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.