Learn More
N,N'-Diphenylthiourea, 98%
Description
N,N'-Diphenylthiourea possesses a wide dipole moment and thus is involved in the forming wide metal chelated complexes as the radioactive-compound which used in radiopharmaceutical imaging, and to treat metal poisoning. It is used in co-crystals development used in the field of nonlinear optics to generate new coherent wavelengths.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N′-Diphenylthiourea possesses a wide dipole moment and thus is involved in the forming wide metal chelated complexes as the radioactive-compound which used in radiopharmaceutical imaging, and to treat metal poisoning. It is used in co-crystals development used in the field of nonlinear optics to generate new coherent wavelengths.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 150°C to 154°C |
| Density | 1.32 |
| Quantity | 2500 g |
| UN Number | UN2811 |
| Beilstein | 644277 |
| Merck Index | 14,3333 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 228.32 |
| Percent Purity | 98% |
| Chemical Name or Material | N,N'-Diphenylthiourea |
Safety and Handling
| Hazard Category | H317-H361d-H373 |
| Hazard Statement | GHS H Statement H300-H317 Fatal if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 203-004-2 |
| RTECSNumber | FE1225000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.