Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Ditridecyl-3,4,9,10-perylenetetracarboxylic Diimide 98.0+%, TCI America™
SDP

Catalog No. D39471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39471G 1 g
D3947200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39471G Supplier TCI America Supplier No. D39471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95689-92-2
Molecular Formula C50H62N2O4
Molecular Weight (g/mol) 755.056
MDL Number MFCD00134587
InChI Key XCAZCFDCJHGAIT-UHFFFAOYSA-N
Synonym PTCDI-C13
PubChem CID 3295215
SMILES CCCCCCCCCCCCCN1C(=O)C2=C3C(=CC=C4C3=C(C=C2)C5=C6C4=CC=C7C6=C(C=C5)C(=O)N(C7=O)CCCCCCCCCCCCC)C1=O

Specifications

Quantity 1 g
Formula Weight 755.06
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Ditridecyl-3,4,9,10-perylenetetracarboxylic Diimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.