Learn More
Sigma Organic Chemistry N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine | 100ML | 140-07-8 | MFCD00002854
Supplier: Sigma Organic Chemistry 87600100ML

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 100ML
About This Item:
Density: 1.1 g/mL at 20 C (lit.)
Refractive Index: n20/D 1.501
Storage: room temp
EINECS Number: 205-396-0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.