Learn More
Sigma Organic Chemistry N,N,N',N'',N''-Pentamethyldiethylenetriamine | 1L | 3030-47-5 | MFCD00014876 | 0.99
Supplier: Sigma Organic Chemistry 3694971L

N,N,N',N'',N''-Pentamethyldiethylenetriamine, 1L
About This Item:
Synonym(s): PMDETA; PMDTA
Linear Formula: [(CH3)2NCH2CH2]2NCH3
Molecular Weight: 173.3
CAS Number: 3030-47-5
MDL Number: MFCD00014876
UNSPSC Code: 12162002
Purity: 0.99
Boiling Point: 198 C (lit.)
Melting Point: -20 C (lit.)
Density: 0.83 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.442 (lit.)
Storage: room temp
EINECS Number: 221-201-1
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.