Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nicotinoylglycine 98.0+%, TCI America™
SDP

Catalog No. N009225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N009225G 25 g
N00925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N009225G Supplier TCI America Supplier No. N009225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 583-08-4
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00023578
InChI Key ZBSGKPYXQINNGF-UHFFFAOYSA-N
Synonym nicotinuric acid, nicotinylglycine, nicotinoylglycine, 2-nicotinamido acetic acid, n-nicotinoylglycine, n-pyridin-3-ylcarbonyl glycine, nicotinoyl-glycine, n-nicotinylglycine, nicotinurate, n-nicotinoyl-glycine
PubChem CID 68499
ChEBI CHEBI:7563
IUPAC Name 2-(pyridine-3-carbonylamino)acetic acid
SMILES C1=CC(=CN=C1)C(=O)NCC(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Nicotinoylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.