Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosodi-n-butylamine Solution, 100μg/mL in Methanol, Ultra Scientific

Catalog No. USNS1301
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-NS-1301 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-NS-1301 Supplier Agilent Technologies Supplier No. NS1301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 924-16-3 , 67-56-1
Molecular Formula C8H18N2O
Molecular Weight (g/mol) 158.245
InChI Key YGJHZCLPZAZIHH-UHFFFAOYSA-N
PubChem CID 13542
ChEBI CHEBI:82356
IUPAC Name N,N-dibutylnitrous amide
SMILES CCCCN(CCCC)N=O

Specifications

CAS Max % 99.99, 0.01
Physical Form Liquid
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material N-Nitrosodi-n-butylamine Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.