Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosodi-n-propylamine Solution, 100μg/mL in Methanol, Ultra Scientific

Catalog No. USNS1201
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-NS-1201 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-NS-1201 Supplier Agilent Technologies Supplier No. NS1201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 621-64-7 , 67-56-1
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
InChI Key YLKFDHTUAUWZPQ-UHFFFAOYSA-N
PubChem CID 12130
ChEBI CHEBI:82358
IUPAC Name N,N-dipropylnitrous amide
SMILES CCCN(CCC)N=O

Specifications

Physical Form Liquid
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material N-Nitrosodi-n-propylamine Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.