Learn More
Abcam N-Nitrosodibenzylamine, Micronucleus-inducing Mitogenic Agent, Solid, 50 mg
Description
- Molecular Weight: 226.27 Da
- Form: Solid
- Activity: Micronucleus-inducing mitogenic agent
- Biological Effects: Mutagenic to Salmonella typhimurium; Genotoxic to rat hepatocytes; Active in vitro and in vivo
- Composition: 100% dibenzyl(nitroso)amine
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | N-Nitrosodibenzylamine |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Buffer | 100% dibenzyl(nitroso)amine |
| Formulation | 100% dibenzyl(nitroso)amine |
| Quantity | 50 mg |
| For Use With (Application) | Micronucleus induction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.