Learn More
AdipoGen N-Nitrosomethylphenylamine
SDP
Supplier:  AdipoGen CDXN0372G001
Encompass_Preferred
Chemical. CAS 614-00-6. Formula C7H8N2O. MW 136.15. N-nitrosomethylphenylamine NMPA is a nitrosamine and is known to be a potent carcinogen associated with cancer development in animals and humans. NMPA can damage DNA and cause mutations in genes that control cell growth and division. It reacts with DNA in vitro to form genotoxic activity, which may lead to cell death or mutagenesis. NMPA is formed in various industrial processes, including the synthesis of certain chemicals. It serves as a building block in the synthesis of diverse compounds and functions as an analytical reagent for environmental monitoring. This compound can be used as analytical reference material. NMPA is useful for studying enzymatic denitrosation processes. NMPA could be used as a tumor initiator, as a reagent for preparing carcinogenic animal diseases model.
Catalog No. 50-256-0360