Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosopiperidine Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. usns1701
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-NS-1701 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-NS-1701 Supplier Agilent Technologies Supplier No. NS1701
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-75-4 , 67-56-1
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.148
InChI Key UWSDONTXWQOZFN-UHFFFAOYSA-N
PubChem CID 7526
ChEBI CHEBI:76324
IUPAC Name 1-nitrosopiperidine
SMILES C1CCN(CC1)N=O

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Methanol,N-Nitrosopiperidine
Element Methanol,N-Nitrosopiperidine
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material N-Nitrosopiperidine Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.