Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Nonylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
112-20-9
C9H21N
143.27
MFCD00008249
FJDUDHYHRVPMJZ-UHFFFAOYSA-N
nonylamine,n-nonylamine,1-aminononane,1-nonanamine,1-nonylamine,unii-7l7h2ht4ok,7l7h2ht4ok,nonyl-amine,n-nonyl amine,amine c9
16215
nonan-1-amine
CCCCCCCCCN
112-20-9
C9H21N
143.27
MFCD00008249
FJDUDHYHRVPMJZ-UHFFFAOYSA-N
nonylamine,n-nonylamine,1-aminononane,1-nonanamine,1-nonylamine,unii-7l7h2ht4ok,7l7h2ht4ok,nonyl-amine,n-nonyl amine,amine c9
16215
nonan-1-amine
CCCCCCCCCN
Specific Gravity 0.784
Refractive Index 1.4320 to 1.4340
Melting Point -1.0°C
Color Colorless to Yellow
Boiling Point 202.0°C
Quantity 25mL
Flash Point 63°C
Packaging Glass bottle
Infrared Spectrum Authentic
Density 0.7840g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3(CH2)8NH2
Beilstein 04,198
Solubility Information Solubility in water: slightly soluble
Formula Weight 143.27
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material n-Nonylamine
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 203-945-9