Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octanal Diethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. O04275ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04275ML 5 mL
O042725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04275ML Supplier TCI America Supplier No. O04275ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54889-48-4
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.338
MDL Number MFCD00672807
InChI Key BRKIPAWFUDCCOO-UHFFFAOYSA-N
Synonym 1,1-Diethoxyoctane, n-Octyl Aldehyde Diethyl Acetal
PubChem CID 101364
IUPAC Name 1,1-diethoxyoctane
SMILES CCCCCCCC(OCC)OCC

Specifications

Color Colorless
Boiling Point 115°C
Quantity 5 mL
UN Number 1993
Formula Weight 202.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octanal Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.