Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octane, 98+%

Catalog No. p-7021018
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13181AE 100 mL
AAA13181AP 500 mL
AAA131810F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA13181AE Supplier Thermo Scientific Chemicals Supplier No. A13181AE
Only null left
Add to Cart

CAS: 111-65-9 | C8H18 | 114.232 g/mol

N-octane is mainly used as a solvent, fuel and as one of the standard values in octane rating scale. It is also used to prevent uneven burning of fuel in internal-combustion engines. Further, it acts as an antiknock agent for high compression engine fuels, lacquer diluents, blowing agent for foam rubber. In addition, it is used in rocket propellants and in azeotropic distillations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-octane is mainly used as a solvent, fuel and as one of the standard values in octane rating scale. It is also used to prevent uneven burning of fuel in internal-combustion engines. Further, it acts as an antiknock agent for high compression engine fuels, lacquer diluents, blowing agent for foam rubber. In addition, it is used in rocket propellants and in azeotropic distillations.

Solubility
Miscible with ethyl ether, ethanol, acetone, benzene, petroleum ether and gasoline. Slightly Miscible with alcohol. Immiscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents and acids
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Specifications

Melting Point -57°C
Density 0.703
Boiling Point 125°C to 126°C
Flash Point 15°C (59°F)
Odor Nearly Odorless
Assay Percent Range ≥98%
Refractive Index 1.398
Quantity 100 mL
UN Number UN1262
Beilstein 1696875
Merck Index 14,6749
Solubility Information Miscible with ethyl ether,ethanol,acetone,benzene,petroleum ether and gasoline; Slightly Miscible with alcohol; Immiscible with water.
Formula Weight 114.23
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material n-Octane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
EINECSNumber 203-892-1
RTECSNumber RG8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.