Learn More
Avanti Polar Lipids N-OCTYL-B-D-GLUCOSIDE 1G
Supplier:  Avanti Polar Lipids 850511P1G
Encompass
n-octyl-ß-D-glucopyranoside.Octyl glucoside is a dialyzable nonionic detergent for functional solubilization and purification of membrane proteins.
Catalog No. NC1736117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.