Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam n-Octyl-beta-D-maltopyranoside, Non-ionic Detergent, >98% Purity, 1 g
Description
- Molecular weight: 454.4 g/mol
- Purity: >98%
- Description: Water soluble non-ionic detergent
- Application: Suitable for solubilization and isolation of membrane proteins
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Format | Bottle |
| Form | Powder |
| Target Molecule | Membrane Proteins |
| Quantity | 1 g |
| For Use With (Application) | Solubilization and isolation of membrane proteins |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?