Learn More
Abcam n-Octyl-beta-D-maltopyranoside, non-ionic detergent, 250MG
Supplier:  Abcam AB146326250MG
Encompass_Preferred
MW 454.4 g/mol, Purity >98%. Water soluble non-ionic detergent suitable for solubilization and isolation of membrane proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4716
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.