Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyl-β-D-Glucopyranoside, MP Biomedicals™

Catalog No. MP215005410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
10 g
25 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215005410 10 g
MP21500542 250 mg
MP21500545 500 mg
ICN15005401 1 g
ICN15005405 5 g
MP215005425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP215005410 Supplier MP Biomedicals, Inc Supplier No. 0215005410
Add to Cart
Add to Cart

n-Octyl-β-D-Glucopyranoside >98%, white powder, non-ionic detergent for membrane protein.

  • Non-ionic alkyl-glucoside detergent commonly used in the solubilization of membrane proteins and phospholipids.
  • Used at a molar dependant ratios for the solubilization of phosphatidylcholine bilayers.
  • Used for protein crystallization, as well as for enhancement of oligosaccharide cleavage from glycoproteins using endoglycosidase F.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym n-Octyl-b-D-glucoside, OGP
PubChem CID 62852
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Physical Form Powder
Melting Point 89°C to 100°C (literature)
Quantity 10 g
Specific Rotation -31 ± 3° (c=5, water)
Beilstein 84118
Solubility Information Soluble in water (10mg/mL-clear colorless solution).
Grade 98%
storageNote1 20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.