Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyltrichlorosilane 98.0+%, TCI America™
SDP

Catalog No. O0168250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0168250G 250 g
O016825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0168250G Supplier TCI America Supplier No. O0168250G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 5283-66-9
Molecular Formula C8H17Cl3Si
Molecular Weight (g/mol) 247.659
MDL Number MFCD00000488
InChI Key RCHUVCPBWWSUMC-UHFFFAOYSA-N
Synonym trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
PubChem CID 21354
IUPAC Name trichloro(octyl)silane
SMILES CCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 229°C
Quantity 250 g
UN Number 1801
Formula Weight 247.66
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material n-Octyltrichlorosilane
EINECSNumber (2)-2041
RTECSNumber VV4700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.